3,4-Dimethoxybenzophenone 98%
Leverandør: Apollo Scientific
|
Warning
|
Synonymer:
(3,4-Dimethoxyphenyl)(phenyl)methanone
OR40183-100G
APOSOR40183-100GEA
1000
NOK
APOSOR40183-100G
3,4-Dimethoxybenzophenone 98%
3,4-Dimethoxybenzophenone
Formel:
C₁₅H₁₄O₃ MW: 242,2700 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00075875 CAS-nummer: 4038-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...