Triphenylene 97%
Leverandør: Apollo Scientific
|
Danger
|
Synonymer:
9,10-Benzophenanthrene, Isochrysene
OR41025-25G
APOSOR41025-25GEA
790
NOK
APOSOR41025-25G
Triphenylene 97%
Triphenylene
Formel:
C₁₈H₁₂ MW: 228,29 g/mol Kokepunkt: 438 °C Smeltepunkt: 195…198 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00001108 CAS-nummer: 217-59-4 EINECS: 205-922-9 FN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...