2,4-Dimethyloxazole-5-carboxylic acid

Leverandør: Apollo Scientific
Warning

OR46096-10G OR46096-1G OR46096-5G
APOSOR46096-10GEA 1700 NOK
APOSOR46096-10G APOSOR46096-1G APOSOR46096-5G
2,4-Dimethyloxazole-5-carboxylic acid
2,4-Dimethyloxazole-5-carboxylic acid
Formel: C₆H₇NO₃
MW: 141,13 g/mol
Smeltepunkt: 242 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06011090
CAS-nummer: 2510-37-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR