(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%

Leverandør: Apollo Scientific
Warning

Synonymer: (3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate

OR4642-5G
APOSOR4642-5GEA 270 NOK
APOSOR4642-5G
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formel: C₉H₁₈N₂O₂
MW: 186,26 g/mol
Smeltepunkt: 50…54 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00143191
CAS-nummer: 122536-77-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR