2-Acetyl-5-chlorothiophene 99%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20

OR4686-100G OR4686-25G OR4686-500G
APOSOR4686-100GEA 1025 NOK
APOSOR4686-100G APOSOR4686-25G APOSOR4686-500G
2-Acetyl-5-chlorothiophene 99%
2-Acetyl-5-chlorothiophene
Formel: C₆H₅ClOS
MW: 160,62 g/mol
Kokepunkt: 117…118 °C/17mmHg
Smeltepunkt: 46…49 °C
Flash punkt: 108 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005444
CAS-nummer: 6310-09-4
EINECS: 228-630-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR