Methyl-2-aminopyrimidine-5-carboxylate 96%
Leverandør: Apollo Scientific
Synonymer:
Methyl 2-aminopyrimidine-5-carboxylate
OR480769-5G
APOSOR480769-5GEA
1475
NOK
APOSOR480769-5G
Methyl-2-aminopyrimidine-5-carboxylate 96%
Methyl-2-aminopyrimidine-5-carboxylate
Formel:
C₆H₇N₃O₂ MW: 153,1400 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD08275693 CAS-nummer: 308348-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...