1-Boc-3,3-dimethylpiperazine 95%
Leverandør: Apollo Scientific
Synonymer:
1-Boc-3,3-Dimethylpiperazine, tert-Butyl 3,3-dimethylpiperazine-1-carboxylate
OR480840-5G
APOSOR480840-5GEA
1475
NOK
APOSOR480840-5G
1-Boc-3,3-dimethylpiperazine 95%
1-Boc-3,3-dimethylpiperazine
Formel:
C₁₁H₂₂N₂O₂ MW: 214,3046 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD07371651 CAS-nummer: 259808-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...