Bis[4-(3-aminophenoxy)phenyl]sulphone

Leverandør: Apollo Scientific

Synonymer: 3,3'-((sulphonylbis(4,1-phenylene))bis(oxy))dianiline

OR49078-50G
APOSOR49078-50GEA 0 NOK
APOSOR49078-50G
Bis[4-(3-aminophenoxy)phenyl]sulphone
Bis[4-(3-aminophenoxy)phenyl]sulphone
Formel: C₂₄H₂₀N₂O₄S
MW: 432,49 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00054738
CAS-nummer: 30203-11-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR