2-(3-Bromophenyl)triphenylene 98%
Leverandør: Apollo Scientific
OR51010-5G
APOSOR51010-5GEA
1000
NOK
APOSOR51010-5G
2-(3-Bromophenyl)triphenylene 98%
2-(3-Bromophenyl)triphenylene
Formel:
C₂₄H₁₅Br MW: 383,29 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD28384139 CAS-nummer: 1313514-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...