(R)-(+)-2-Chloropropionic acid 98%

Leverandør: Apollo Scientific
Danger

Synonymer: (R)-(+)-2-Chloropropanoic acid

OR52411-25G OR52411-5G OR52411-100G
APOSOR52411-25GEA 610 NOK
APOSOR52411-25G APOSOR52411-5G APOSOR52411-100G
(R)-(+)-2-Chloropropionic acid 98%
(R)-(+)-2-Chloropropionic acid
Formel: H₃CCHClCOOH
MW: 108,52 g/mol
Smeltepunkt: –12…0 °C
Tetthet: 1,258 g/cm³ (25 °C)
Flash punkt: 60 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064204
CAS-nummer: 7474-05-7
FN: 2511
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR