2-klortoluen 99%

Leverandør: Apollo Scientific
Warning

Synonymer: o-Chlorotoluene

OR5337-1KG OR5337-250G OR5337-5KG
APOSOR5337-1KGEA 450 NOK
APOSOR5337-1KG APOSOR5337-250G APOSOR5337-5KG
2-klortoluen 99%
2-klortoluen
Formel: C₇H₇Cl
MW: 126,59 g/mol
Kokepunkt: 157…159 °C
Smeltepunkt: –36 °C
Tetthet: 1,082 g/cm³ (25 °C)
Flash punkt: 47 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000562
CAS-nummer: 95-49-8
EINECS: 202-424-3
FN: 2238
ADR: 3,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR