Methyl-o-toluate 98%
Leverandør: Apollo Scientific
Warning
Synonymer: Methyl 2-methylbenzoate, o-Toluic acid methyl ester, 2-Toluic acid methyl ester, Methyl-2-methylbenzoate, 2-Methylbenzoic acid methyl ester
OR5438-100G OR5438-25G OR5438-500G
APOSOR5438-100GEA 450 NOK
APOSOR5438-100G APOSOR5438-25G APOSOR5438-500G
Methyl-o-toluate 98%
Methyl-o-toluate
Formel: C₉H₁₀O₂
MW: 150,18 g/mol
Kokepunkt: 207…208 °C
Tetthet: 1,072 g/cm³ (25 °C)
Flash punkt: 82 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008428
CAS-nummer: 89-71-4
EINECS: 201-932-2