Methyl-2-chloro-4-thiazolecarboxylate 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Methyl2-chloro-4-thiazolecarboxylate

OR5610-1G OR5610-5G
APOSOR5610-1GEA 325 NOK
APOSOR5610-1G APOSOR5610-5G
Methyl-2-chloro-4-thiazolecarboxylate 98%
Methyl-2-chloro-4-thiazolecarboxylate
Formel: C₅H₄ClNO₂S
MW: 177,6088 g/mol
Smeltepunkt: 90…94 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06659909
CAS-nummer: 850429-61-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR