6-Bromonicotinonitrile 95%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Bromo-5-cyanopyridine

OR5716-25G OR5716-5G
APOSOR5716-25GEA 895 NOK
APOSOR5716-25G APOSOR5716-5G
6-Bromonicotinonitrile 95%
6-Bromonicotinonitrile
Formel: C₆H₃BrN₂
MW: 183,01 g/mol
Smeltepunkt: 143…147 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00234141
CAS-nummer: 139585-70-9
FN: 3439
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR