2-Methyl-5-thiazolecarboxylic acid
Leverandør: Apollo Scientific
|
Warning
|
Synonymer:
2-Methylthiazole-5-carboxylic acid
OR59883-1G
OR59883-250MG
APOSOR59883-1GEA
270
NOK
APOSOR59883-1G
APOSOR59883-250MG
2-Methyl-5-thiazolecarboxylic acid
2-Methyl-5-thiazolecarboxylic acid
Formel:
C₅H₅NO₂S MW: 143,1637 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD01760300 CAS-nummer: 40004-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...