1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride

Leverandør: Apollo Scientific
Warning

Synonymer: O-(4-Chlorobenzyl)hydroxylamine hydrochloride

OR6060-1G OR6060-5G
APOSOR6060-1GEA 2075 NOK
APOSOR6060-1G APOSOR6060-5G
1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride
1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride
Formel: C₇H₉Cl₂NO
MW: 194,0585 g/mol
Smeltepunkt: 230 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01114589
CAS-nummer: 38936-60-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR