2-Sulphobenzoic acid 95%
Leverandør: Apollo Scientific
|
Danger
|
OR61312-100G
OR61312-25G
OR61312-5G
APOSOR61312-100GEA
12950
NOK
APOSOR61312-100G
APOSOR61312-25G
APOSOR61312-5G
2-Sulphobenzoic acid 95%
2-Sulphobenzoic acid
Formel:
C₇H₆O₅S Smeltepunkt: 142 °C Lagringstemperatur: Romtemperatur |
CAS-nummer:
632-25-7 FN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...