2-Chloro-5-pyridineboronic acid pinacol ester 97%
Leverandør: Apollo Scientific
|
Warning
|
Synonymer:
2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR7183-5G
APOSOR7183-5GEA
485
NOK
APOSOR7183-5G
2-Chloro-5-pyridineboronic acid pinacol ester 97%
2-Chloro-5-pyridineboronic acid pinacol ester
Formel:
C₁₁H₁₅BClNO₂ MW: 239,51 g/mol Smeltepunkt: 86…90 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD03412793 CAS-nummer: 444120-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...