4-(3-Bromophenyl)morpholine 96%
Leverandør: Apollo Scientific
|
Warning
|
OR8531-1G
OR8531-5G
OR8531-25G
APOSOR8531-1GEA
270
NOK
APOSOR8531-1G
APOSOR8531-5G
APOSOR8531-25G
4-(3-Bromophenyl)morpholine 96%
4-(3-Bromophenyl)morpholine
Formel:
C₁₀H₁₂BrNO MW: 242,1100 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD04112585 CAS-nummer: 197846-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...