Ethyl-5-chloro-1-methyl-1H-pyrazole-3-carboxylate 95%
Leverandør: Apollo Scientific
Synonymer:
Ethyl 5-chloro-1-methyl-1H-pyrazole-3-carboxylate
OR903729-1G
OR903729-250MG
APOSOR903729-1GEA
5775
NOK
APOSOR903729-1G
APOSOR903729-250MG
Ethyl-5-chloro-1-methyl-1H-pyrazole-3-carboxylate 95%
Ethyl-5-chloro-1-methyl-1H-pyrazole-3-carboxylate
Formel:
C₇H₉ClN₂O₂ MW: 188,61 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD10699191 CAS-nummer: 1172229-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...