2-Hydrazinobenzothiazole 97%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Hydrazinylbenzo[d]thiazole

OR9046-10G OR9046-50G OR9046-25G
APOSOR9046-10GEA 0 NOK
APOSOR9046-10G APOSOR9046-50G APOSOR9046-25G
2-Hydrazinobenzothiazole 97%
2-Hydrazinobenzothiazole
Formel: C₇H₇N₃S
MW: 165,21 g/mol
Smeltepunkt: 198…202 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00041849
CAS-nummer: 615-21-4
EINECS: 210-416-6
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR