4'-Methylformanilide 95%

Leverandør: Apollo Scientific

OR908773-1G OR908773-5G
APOSOR908773-1GEA 2075 NOK
APOSOR908773-1G APOSOR908773-5G
4'-Methylformanilide 95%
4'-Methylformanilide
Formel: C₈H₉NO
Lagringstemperatur: Romtemperatur
CAS-nummer: 3085-54-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR