4-tert-Butylbensosyre 95%

Leverandør: Apollo Scientific

Synonymer: 4-(tert-Butyl)benzoic acid

OR911294-1KG
APOSOR911294-1KGEA 0 NOK
APOSOR911294-1KG
4-tert-Butylbensosyre 95%
4-tert-Butylbensosyre
Formel: C₁₁H₁₄O₂
MW: 178,23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002563
CAS-nummer: 98-73-7
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR