2-Pyrazinecarbonyl chloride 95%

Leverandør: Apollo Scientific

Synonymer: Pyrazine-2-carbonyl chloride

OR912568-1G
APOSOR912568-1GEA 4900 NOK
APOSOR912568-1G
2-Pyrazinecarbonyl chloride 95%
2-Pyrazinecarbonyl chloride
Formel: C₅H₃ClN₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00792886
CAS-nummer: 19847-10-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR