1-(tert-Butoxycarbonyl)-3-iodopyrrolidine 97%

Leverandør: Apollo Scientific

Synonymer: 1-Boc-3-iodo-pyrrolidine, tert-Butyl 3-iodopyrrolidine-1-carboxylate

OR919042-1G
APOSOR919042-1GEA 965 NOK
APOSOR919042-1G
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine 97%
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine
Formel: C₉H₁₆INO₂
MW: 297,14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12031283
CAS-nummer: 774234-25-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR