5-Methyl-1H-pyrazole-3-carbonitrile 98%

Leverandør: Apollo Scientific

OR926328-1G OR926328-250MG OR926328-5G
APOSOR926328-1GEA 1500 NOK
APOSOR926328-1G APOSOR926328-250MG APOSOR926328-5G
5-Methyl-1H-pyrazole-3-carbonitrile 98%
5-Methyl-1H-pyrazole-3-carbonitrile
Formel: C₅H₅N₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11934282
CAS-nummer: 38693-82-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR