2-(Phenylthio)acetamide 95%

Leverandør: Apollo Scientific

OR927470-1G OR927470-5G
APOSOR927470-1GEA 2350 NOK
APOSOR927470-1G APOSOR927470-5G
2-(Phenylthio)acetamide 95%
2-(Phenylthio)acetamide
Formel: C₈H₉NOS
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00040684
CAS-nummer: 22446-20-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR