[5-(4-Methylphenyl)isoxazol-3-yl]methanol 95%

Leverandør: Apollo Scientific

OR930054-1G OR930054-250MG
APOSOR930054-1GEA 9925 NOK
APOSOR930054-1G APOSOR930054-250MG
[5-(4-Methylphenyl)isoxazol-3-yl]methanol 95%
[5-(4-Methylphenyl)isoxazol-3-yl]methanol
Formel: C₁₁H₁₁NO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06199339
CAS-nummer: 640291-93-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR