3-Bromo-2-nitrotoluene 97+%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-Bromo-3-methyl-2-nitrobenzene

OR930058-25G OR930058-5G
APOSOR930058-25GEA 1350 NOK
APOSOR930058-25G APOSOR930058-5G
3-Bromo-2-nitrotoluene 97+%
3-Bromo-2-nitrotoluene
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
Smeltepunkt: 25…29 °C
Tetthet: 1,612 g/cm³ (25 °C)
Flash punkt: >230 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02673631
CAS-nummer: 52414-97-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR