2-Chloro-4-phenylpyrimidine 95%
Leverandør: Apollo Scientific
|
Danger
|
OR9304-1G
APOSOR9304-1GEA
415
NOK
APOSOR9304-1G
2-Chloro-4-phenylpyrimidine 95%
2-Chloro-4-phenylpyrimidine
Formel:
C₁₀H₇ClN₂ Smeltepunkt: 84…88 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00234600 CAS-nummer: 13036-50-5 FN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...