9-Fluorenone-2,7-dicarboxylic acid, Technical Grade
Leverandør: Apollo Scientific
Synonymer: 9-Oxo-9H-fluorene-2,7-dicarboxylic acid
OR930734-1G OR930734-5G
APOSOR930734-1GEA 750 NOK
APOSOR930734-1G APOSOR930734-5G
9-Fluorenone-2,7-dicarboxylic acid, Technical Grade
9-Fluorenone-2,7-dicarboxylic acid
Formel: C₁₅H₈O₅
MW: 268,23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016355
CAS-nummer: 792-26-7