3-Methyl-N-methylbenzylamine 95%

Leverandør: Apollo Scientific

Synonymer: N-Methyl-1-(m-tolyl)methanamine

OR936302-5G
APOSOR936302-5GEA 2325 NOK
APOSOR936302-5G
3-Methyl-N-methylbenzylamine 95%
3-Methyl-N-methylbenzylamine
Formel: C₉H₁₃N
MW: 135,21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05865181
CAS-nummer: 39180-84-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR