3-(4-Hydroxyphenyl)benzonitrile 98%
Leverandør: Apollo Scientific
Synonymer:
4'-Hydroxy-[1,1'-biphenyl]-3-carbonitrile, 4-(3-Cyanophenyl)phenol
OR954778-5G
APOSOR954778-5GEA
4100
NOK
APOSOR954778-5G
3-(4-Hydroxyphenyl)benzonitrile 98%
3-(4-Hydroxyphenyl)benzonitrile
Formel:
C₁₃H₉NO MW: 195,2200 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD05854255 CAS-nummer: 154848-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...