5-Ethoxy-2-pyridine carboxylic acid 95%
Leverandør: Apollo Scientific
Synonymer:
5-Ethoxypicolinic acid
OR960468-1G
OR960468-250MG
APOSOR960468-1GEA
1975
NOK
APOSOR960468-1G
APOSOR960468-250MG
5-Ethoxy-2-pyridine carboxylic acid 95%
5-Ethoxy-2-pyridine carboxylic acid
Formel:
C₈H₉NO₃ MW: 167,17 g/mol |
MDL nummer:
MFCD13562922 CAS-nummer: 98353-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...