(R)-(-)-α-Methoxyphenylacetic acid 98%

Leverandør: Apollo Scientific

OR960661-100G OR960661-25G
APOSOR960661-100GEA 3975 NOK
APOSOR960661-100G APOSOR960661-25G
(R)-(-)-α-Methoxyphenylacetic acid 98%
(R)-(-)-α-Methoxyphenylacetic acid
Formel: C₉H₁₀O₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004250
CAS-nummer: 3966-32-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR