1-Chloro-3-methylisoquinoline 97%
Leverandør: Apollo Scientific
OR961835-1G
OR961835-250MG
APOSOR961835-1GEA
9325
NOK
APOSOR961835-1G
APOSOR961835-250MG
1-Chloro-3-methylisoquinoline 97%
1-Chloro-3-methylisoquinoline
Formel:
C₁₀H₈ClN Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD09261357 CAS-nummer: 7115-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...