C-(5-Phenyl[1,3,4]oxadiazol-2-yl)methylamine 95%

Leverandør: Apollo Scientific

OR962280-1G OR962280-5G
APOSOR962280-1GEA 1625 NOK
APOSOR962280-1G APOSOR962280-5G
C-(5-Phenyl[1,3,4]oxadiazol-2-yl)methylamine 95%
C-(5-Phenyl[1,3,4]oxadiazol-2-yl)methylamine
Formel: C₉H₉N₃O
MW: 175,19 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06490519
CAS-nummer: 46182-58-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR