2-Amino-7,8-dihydroquinazolin-5(6H)-one hydrochloride 95%
Leverandør: Apollo Scientific
Synonymer:
2-Amino-7,8-dihydroquinazolin-5(6h)-one
OR963736-1G
OR963736-250MG
APOSOR963736-1GEA
2500
NOK
APOSOR963736-1G
APOSOR963736-250MG
2-Amino-7,8-dihydroquinazolin-5(6H)-one hydrochloride 95%
2-Amino-7,8-dihydroquinazolin-5(6H)-one hydrochloride
Formel:
C₈H₁₀ClN₃O MW: 199,64 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD20526329 CAS-nummer: 21599-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...