2-Methoxy-4-nitrobenzaldehyde 95%
Leverandør: Apollo Scientific
OR963995-5G
APOSOR963995-5GEA
2625
NOK
APOSOR963995-5G
2-Methoxy-4-nitrobenzaldehyde 95%
2-Methoxy-4-nitrobenzaldehyde
Formel:
C₈H₇NO₄ MW: 181,15 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD02683052 CAS-nummer: 136507-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...