2-Amino-3-bromophenol 95%
Leverandør: Apollo Scientific
|
Warning
|
OR9739-1G
OR9739-250MG
OR9739-5G
APOSOR9739-1GEA
715
NOK
APOSOR9739-1G
APOSOR9739-250MG
APOSOR9739-5G
2-Amino-3-bromophenol 95%
2-Amino-3-bromophenol
Formel:
C₆H₆BrNO MW: 188,0200 g/mol Smeltepunkt: 135…137 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD08445676 CAS-nummer: 116435-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...