1,3,5-Triklorbensen

Leverandør: Apollo Scientific
Warning

OR9872-100G OR9872-1KG OR9872-500G
APOSOR9872-100GEA 255 NOK
APOSOR9872-100G APOSOR9872-1KG APOSOR9872-500G
1,3,5-Triklorbensen
1,3,5-Triklorbensen
Formel: C₆H₃Cl₃
MW: 181,45 g/mol
Kokepunkt: 208 °C
Smeltepunkt: 56…65 °C
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000585
CAS-nummer: 108-70-3
EINECS: 203-608-6
FN: 3077
ADR: 9+EHS,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR