Ethyl iodofluoroacetate 98%

Leverandør: Apollo Scientific
Warning

PC0896-25G PC0896-5G
APOSPC0896-25GEA 4125 NOK
APOSPC0896-25G APOSPC0896-5G
Ethyl iodofluoroacetate 98%
Ethyl iodofluoroacetate
Formel: C₄H₆FIO₂
Kokepunkt: 43 °C/3mmHg
Tetthet: >1,4 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01320733
CAS-nummer: 401-58-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR