2,3,4-Trifluorophenylboronic acid

Leverandør: Apollo Scientific
Warning

PC10547-100G PC10547-25G PC10547-5G
APOSPC10547-100GEA 2475 NOK
APOSPC10547-100G APOSPC10547-25G APOSPC10547-5G
2,3,4-Trifluorophenylboronic acid
2,3,4-Trifluorophenylboronic acid
Formel: C₆H₄BF₃O₂
MW: 175,9 g/mol
Smeltepunkt: 229…235 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01863168
CAS-nummer: 226396-32-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR