Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate
Leverandør: Apollo Scientific
PC200021-1G
PC200021-500MG
APOSPC200021-1GEA
3850
NOK
APOSPC200021-1G
APOSPC200021-500MG
Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate
Methyl-3-{4-[(2-bromo-5-fluorophenyl)methoxy]phenyl}propanoate
Formel:
C₁₇H₁₆BrFO₃ MW: 367,22 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD22573516 CAS-nummer: 1407516-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...