1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

Leverandør: Apollo Scientific

PC200289-1G PC200289-500MG
APOSPC200289-1GEA 7700 NOK
APOSPC200289-1G APOSPC200289-500MG
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
Formel: C₆H₄F₃N₃
Smeltepunkt: 160 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12403326
CAS-nummer: 1049772-79-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR