3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Leverandør: Apollo Scientific
PC200310-500MG
APOSPC200310-500MGEA
5825
NOK
APOSPC200310-500MG
3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel:
C₈H₄F₃IN₂ MW: 312,03 g/mol Smeltepunkt: 205…207 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD11857744 CAS-nummer: 1142192-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...