Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate 98+%
Leverandør: Apollo Scientific
Warning
Synonymer: Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
PC2199-1G PC2199-250MG
APOSPC2199-1GEA 4350 NOK
APOSPC2199-1G APOSPC2199-250MG
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate 98+%
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Formel: C₁₇H₂₄F₃O₃RhS
MW: 468,34 g/mol
Smeltepunkt: 159 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00143752
CAS-nummer: 99326-34-8