3,4-Difluorbenso syre 98%
Leverandør: Apollo Scientific
|
Warning
|
PC2662-100G
PC2662-25G
PC2662-500G
APOSPC2662-100GEA
520
NOK
APOSPC2662-100G
APOSPC2662-25G
APOSPC2662-500G
3,4-Difluorbenso syre 98%
3,4-Difluorbenso syre
Formel:
C₇H₄F₂O₂ MW: 158,1 g/mol Smeltepunkt: 120…122 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00011672 CAS-nummer: 455-86-7 EINECS: 207-249-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...