2-(3,4,5-Trifluorophenyl)ethanol

Leverandør: Apollo Scientific

PC302003-1G
APOSPC302003-1GEA 1850 NOK
APOSPC302003-1G
2-(3,4,5-Trifluorophenyl)ethanol
2-(3,4,5-Trifluorophenyl)ethanol
Formel: C₈H₇F₃O
MW: 176,14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06660355
CAS-nummer: 886761-77-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR