4-Chloro-2-(trifluoromethyl)benzylamine 97%
Leverandør: Apollo Scientific
PC303078-5G
APOSPC303078-5GEA
965
NOK
APOSPC303078-5G
4-Chloro-2-(trifluoromethyl)benzylamine 97%
4-Chloro-2-(trifluoromethyl)benzylamine
Formel:
C₈H₇ClF₃N MW: 209,6 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD06213291 CAS-nummer: 771583-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...